siRNA Details
Gene Symbol: TYMSOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012716.2 | NP_001012734.2 | 1 | 292 | ||
GenBank mRNA | AK289986.1 | - | 1 | 221 | ||
BC028301.1 | - | 1 | 292 | 5141042 | ||
BM695182.1 | - | 1 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
