siRNA Details
Gene Symbol: TYMSOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012716.2 | NP_001012734.2 | 2 | 620 | ||
GenBank mRNA | BC028301.1 | - | 2 | 620 | 5141042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
