siRNA Details
Gene Symbol: CPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080513.3 | NP_001073982.3 | 2 | 2350 | ||
NM_001291988.1 | NP_001278917.1 | 2 | 2347 | |||
XM_005269280.4 | XP_005269337.1 | 3 | 2479 | |||
GenBank mRNA | BC031569.1 | - | 2 | 2298 | 5188827 | |
BC042334.1 | - | 2 | 2306 | 5190234 | ||
BC137398.1 | - | 2 | 2340 | 9021396 | ||
BC137403.1 | - | 2 | 2340 | 9021401 | ||
J05158.2 | - | 1 | 2264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
