siRNA Details
Gene Symbol: C10orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135196.1 | NP_001128668.1 | 3 | 3404 | ||
XM_005269476.4 | XP_005269533.1 | 3 | 3469 | |||
XM_005269477.3 | XP_005269534.1 | 3 | 3389 | |||
XM_005269478.4 | XP_005269535.1 | 3 | 3457 | |||
XM_017015619.1 | XP_016871108.1 | 4 | 3179 | |||
GenBank mRNA | AJ311357.1 | - | 1 | 490 | ||
AJ311358.1 | - | 2 | 1278 | |||
AK056949.1 | - | 1 | 312 | |||
AL833265.1 | - | 3 | 3353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
