siRNA Details
Gene Symbol: ERICH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534735.2 | XP_011533037.1 | 6 | 2720 | ||
XM_017013127.1 | XP_016868616.1 | 8 | 1943 | |||
XM_017013132.1 | XP_016868621.1 | 7 | 1745 | |||
GenBank mRNA | AK090404.1 | - | 3 | 4885 | ||
AK125470.1 | - | 3 | 1632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
