siRNA Details
Gene Symbol: TGFBRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142621.2 | NP_001136093.1 | 12 | 4471 | ||
NM_004257.5 | NP_004248.2 | 12 | 4474 | |||
XM_017005339.1 | XP_016860828.1 | 11 | 3967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
