siRNA Details
Gene Symbol: LOC100129434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125368.1 | 2 | 115 | |||
GenBank mRNA | AK096196.1 | - | 2 | 327 | ||
AK295617.1 | - | 3 | 892 | |||
AK311113.1 | - | 2 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
