siRNA Details
Gene Symbol: ZNF404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033719.2 | NP_001028891.2 | 2 | 200 | ||
GenBank mRNA | AK300120.1 | - | 2 | 244 | ||
AY166785.1 | - | 2 | 144 | |||
BC101332.1 | - | 2 | 194 | |||
BC101333.1 | - | 2 | 200 | 40025019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
