siRNA Details
Gene Symbol: ZNF283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297752.1 | NP_001284681.1 | 6 | 723 | ||
NM_181845.1 | NP_862828.1 | 7 | 778 | |||
XM_005258784.3 | XP_005258841.1 | 7 | 891 | |||
XM_005258785.3 | XP_005258842.1 | 6 | 685 | |||
XM_005258786.3 | XP_005258843.1 | 7 | 850 | |||
XM_017026628.1 | XP_016882117.1 | 8 | 972 | |||
XM_017026629.1 | XP_016882118.1 | 8 | 1013 | |||
XM_017026630.1 | XP_016882119.1 | 8 | 976 | |||
XM_017026631.1 | XP_016882120.1 | 8 | 843 | |||
XM_017026632.1 | XP_016882121.1 | 8 | 1019 | |||
XM_017026633.1 | XP_016882122.1 | 8 | 968 | |||
XM_017026634.1 | XP_016882123.1 | 7 | 721 | |||
XM_017026635.1 | XP_016882124.1 | 6 | 786 | |||
XM_017026636.1 | XP_016882125.1 | 2 | 6491 | |||
XM_017026637.1 | XP_016882126.1 | 6 | 764 | |||
XM_017026638.1 | XP_016882127.1 | 5 | 558 | |||
XM_017026639.1 | XP_016882128.1 | 7 | 886 | |||
GenBank mRNA | AK098175.1 | - | 6 | 635 | ||
AK294852.1 | - | 7 | 801 | |||
AY166784.1 | - | 2 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
