siRNA Details
Gene Symbol: ZNF283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297752.1 | NP_001284681.1 | 6 | 638 | ||
NM_181845.1 | NP_862828.1 | 7 | 693 | |||
XM_005258784.3 | XP_005258841.1 | 7 | 806 | |||
XM_005258785.3 | XP_005258842.1 | 6 | 600 | |||
XM_005258786.3 | XP_005258843.1 | 7 | 765 | |||
XM_017026628.1 | XP_016882117.1 | 8 | 887 | |||
XM_017026629.1 | XP_016882118.1 | 8 | 928 | |||
XM_017026630.1 | XP_016882119.1 | 8 | 891 | |||
XM_017026631.1 | XP_016882120.1 | 8 | 758 | |||
XM_017026632.1 | XP_016882121.1 | 8 | 934 | |||
XM_017026633.1 | XP_016882122.1 | 8 | 883 | |||
XM_017026634.1 | XP_016882123.1 | 7 | 636 | |||
XM_017026635.1 | XP_016882124.1 | 6 | 701 | |||
XM_017026636.1 | XP_016882125.1 | 2 | 6406 | |||
XM_017026637.1 | XP_016882126.1 | 6 | 679 | |||
XM_017026638.1 | XP_016882127.1 | 5 | 473 | |||
XM_017026639.1 | XP_016882128.1 | 7 | 801 | |||
GenBank mRNA | AY166784.1 | - | 2 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
