siRNA Details
Gene Symbol: MYO5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080467.2 | NP_001073936.1 | 40 | 5730 | ||
GenBank mRNA | AB032945.2 | - | 40 | 5460 | ||
AB290160.1 | - | 40 | 5550 | |||
AK025336.1 | - | 19 | 2795 | |||
AK093149.1 | - | 18 | 2410 | |||
AK301976.1 | - | 12 | 1881 | |||
AL133643.1 | - | 18 | 2429 | |||
AY274809.1 | - | 18 | 2409 | |||
BC033527.1 | - | 9 | 1368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
