siRNA Details
Gene Symbol: C18orf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026109.1 | XP_016881598.1 | 1 | 61 | ||
GenBank mRNA | AB027121.1 | - | 1 | 61 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
