siRNA Details
Gene Symbol: LETMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243689.1 | NP_001230618.1 | 3 | 372 | ||
NM_001300765.1 | NP_001287694.1 | 3 | 372 | |||
NM_015416.4 | NP_056231.3 | 3 | 372 | |||
NR_045017.1 | 3 | 247 | ||||
XM_006719339.1 | XP_006719402.1 | 3 | 344 | |||
XM_011538161.2 | XP_011536463.1 | 3 | 379 | |||
XM_011538162.2 | XP_011536464.1 | 3 | 380 | |||
XM_011538163.2 | XP_011536465.1 | 3 | 381 | |||
XM_011538165.2 | XP_011536467.1 | 3 | 379 | |||
XM_017019151.1 | XP_016874640.1 | 3 | 381 | |||
XM_017019152.1 | XP_016874641.1 | 3 | 347 | |||
XM_017019155.1 | XP_016874644.1 | 3 | 219 | |||
XM_017019159.1 | XP_016874648.1 | 3 | 382 | |||
XM_017019160.1 | XP_016874649.1 | 3 | 344 | |||
XM_017019165.1 | XP_016874654.1 | 3 | 344 | |||
XM_017019167.1 | XP_016874656.1 | 3 | 344 | |||
XM_017019168.1 | XP_016874657.1 | 3 | 344 | |||
GenBank mRNA | AF195651.2 | - | 3 | 344 | ||
AF315598.1 | - | 3 | 208 | |||
AK000639.1 | - | 3 | 359 | |||
AK057866.1 | - | 3 | 317 | |||
AK127540.1 | - | 3 | 333 | |||
AK225836.1 | - | 3 | 330 | |||
AK301955.1 | - | 3 | 247 | |||
AK315119.1 | - | 3 | 339 | |||
BC064943.1 | - | 3 | 318 | 6009854 | ||
BX376670.2 | - | 3 | 313 | |||
DC357852.1 | - | 3 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
