siRNA Details
Gene Symbol: PPP1R42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013626.2 | NP_001013648.1 | 3 | 417 | ||
GenBank mRNA | BC055413.1 | - | 3 | 419 | 6614745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
