siRNA Details
Gene Symbol: OR7E156P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002171.2 | 4 | 2493 | |||
GenBank mRNA | AK097469.1 | - | 5 | 2416 | ||
AK097490.1 | - | 4 | 1732 | |||
AK097573.1 | - | 4 | 1771 |
Gene Symbol: OR7E104P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL137719.1 | - | 1 | 650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
