siRNA Details
Gene Symbol: SPATA31C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145124.1 | NP_001138596.1 | 4 | 3214 | ||
XM_011518701.2 | XP_011517003.1 | 5 | 3852 | |||
XM_011518702.1 | XP_011517004.1 | 4 | 3771 | |||
XM_011518703.1 | XP_011517005.1 | 4 | 3651 |
Gene Symbol: SPATA31C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166137.1 | NP_001159609.1 | 4 | 3196 | ||
GenBank mRNA | AK302448.1 | - | 4 | 3196 | ||
AL080179.1 | - | 1 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
