siRNA Details
Gene Symbol: MRVI1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034093.2 | 2 | 327 | |||
NR_034094.2 | 2 | 327 | ||||
NR_046374.1 | 4 | 566 | ||||
NR_046375.1 | 3 | 466 | ||||
GenBank mRNA | BC028123.1 | - | 2 | 291 | 5167345 | |
BI520309.1 | - | 4 | 505 | |||
BU569149.1 | - | 3 | 393 | |||
BU570065.1 | - | 2 | 235 | |||
DB071465.1 | - | 2 | 330 | |||
DB454301.1 | - | 2 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
