siRNA Details
Gene Symbol: MRVI1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034093.2 | 3 | 602 | |||
NR_034094.2 | 3 | 564 | ||||
NR_046374.1 | 5 | 841 | ||||
NR_046375.1 | 4 | 741 | ||||
GenBank mRNA | BC028123.1 | - | 3 | 528 | 5167345 | |
DB071465.1 | - | 4 | 526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
