siRNA Details
Gene Symbol: VPS37D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077621.1 | NP_001071089.1 | 4 | 1398 | ||
XM_017011779.1 | XP_016867268.1 | 4 | 1588 | |||
GenBank mRNA | AY081952.1 | - | 4 | 1398 | ||
BC064621.1 | - | 4 | 1245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
