siRNA Details
Gene Symbol: PKI55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037701.1 | 1 | 2285 | |||
GenBank mRNA | AK125369.1 | - | 1 | 2129 | ||
AL137534.1 | - | 1 | 1589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
