siRNA Details
Gene Symbol: CDC14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003595.1 | 1 | 1738 | |||
GenBank mRNA | BC028690.2 | - | 1 | 1726 | ||
BC068452.1 | - | 1 | 1738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
