siRNA Details
Gene Symbol: LOC541472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131935.1 | 2 | 755 | |||
GenBank mRNA | BC027978.1 | - | 2 | 761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
