siRNA Details
Gene Symbol: FER1L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_119376.1 | 12 | 1061 | |||
GenBank mRNA | AY461812.1 | - | 11 | 1097 | ||
BC029951.1 | - | 12 | 987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
