siRNA Details
Gene Symbol: LINC00921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033904.1 | 1 | 78 | |||
GenBank mRNA | AK096958.1 | - | 1 | 44 | ||
BC104941.1 | - | 1 | 103 | |||
BC104943.1 | - | 1 | 103 | |||
DA532447.1 | - | 1 | 80 |
Gene Symbol: ZNF263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295350.1 | - | 1 | 46 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
