siRNA Details
Gene Symbol: LINC00921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033904.1 | 1 | 1161 | |||
GenBank mRNA | AK096958.1 | - | 1 | 1127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
