siRNA Details
Gene Symbol: CCDC85C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144995.1 | NP_001138467.1 | 6 | 4541 | ||
XM_011536705.2 | XP_011535007.1 | 7 | 4224 | |||
GenBank mRNA | AK026211.1 | - | 1 | 172 | ||
AK093969.1 | - | 1 | 2681 | |||
AK096927.1 | - | 1 | 1329 | |||
AK123840.1 | - | 5 | 3427 | |||
BC029998.1 | - | 1 | 1644 | |||
BC110042.1 | - | 1 | 2804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
