siRNA Details
Gene Symbol: C14orf28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017923.1 | NP_001017923.1 | 4, 5 | 1009 | ||
XM_011536408.1 | XP_011534710.1 | 3, 4 | 310 | |||
GenBank mRNA | AF250395.1 | - | 4, 5 | 993 | ||
BC030258.1 | - | 3, 4 | 747 | |||
BC105030.1 | - | 4, 5 | 1002 | 8144033 | ||
BC105032.1 | - | 4, 5 | 1002 | 8144035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
