siRNA Details
Gene Symbol: C14orf28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017923.1 | NP_001017923.1 | 2 | 625 | ||
GenBank mRNA | AF250395.1 | - | 2 | 609 | ||
AK126985.1 | - | 1 | 381 | |||
BC030258.1 | - | 1 | 363 | |||
BC105030.1 | - | 2 | 618 | 8144033 | ||
BC105032.1 | - | 2 | 618 | 8144035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
