siRNA Details
Gene Symbol: MAPT-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024559.1 | 2 | 176 | |||
GenBank mRNA | AW770488.1 | - | 2 | 176 | ||
BM714794.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
