siRNA Details
Gene Symbol: FAR2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026758.2 | 6 | 2316 | |||
GenBank mRNA | AK125921.1 | - | 6 | 2318 | ||
AL043002.1 | - | 3 | 341 | |||
BC132694.1 | - | 1 | 14 | |||
BC132696.1 | - | 1 | 14 | |||
BC144438.1 | - | 1 | 14 | 9052961 |
Gene Symbol: CYP4F30P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093281.1 | - | 4 | 1473 |
Gene Symbol: LOC440910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030728.1 | 4 | 529 | |||
GenBank mRNA | AK091798.1 | - | 4 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
