siRNA Details
Gene Symbol: FAR2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046258.1 | 6 | 1752 | |||
NR_046259.1 | 5 | 1654 | ||||
NR_046260.1 | 4 | 1529 | ||||
GenBank mRNA | AK054990.1 | - | 4 | 969 | ||
BC131487.1 | - | 2 | 3577 |
Gene Symbol: FAR2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026758.2 | 12 | 3429 | |||
GenBank mRNA | AK125921.1 | - | 12 | 3430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
