siRNA Details
Gene Symbol: PACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100913.2 | NP_001094383.2 | 20 | 2569 | ||
NM_001243127.2 | NP_001230056.1 | 19 | 2046 | |||
NM_015197.3 | NP_056012.2 | 19 | 2524 | |||
XM_006720090.1 | XP_006720153.1 | 20 | 2237 | |||
XM_006720091.1 | XP_006720154.1 | 19 | 2204 | |||
XM_006720092.2 | XP_006720155.1 | 19 | 2165 | |||
XM_017021105.1 | XP_016876594.1 | 20 | 2225 | |||
XM_017021106.1 | XP_016876595.1 | 20 | 2222 | |||
XM_017021107.1 | XP_016876596.1 | 20 | 2213 | |||
XM_017021108.1 | XP_016876597.1 | 20 | 2210 | |||
XM_017021109.1 | XP_016876598.1 | 19 | 2201 | |||
XM_017021110.1 | XP_016876599.1 | 19 | 2192 | |||
XM_017021111.1 | XP_016876600.1 | 19 | 2070 | |||
XM_017021112.1 | XP_016876601.1 | 20 | 2005 | |||
GenBank mRNA | AB011174.1 | - | 20 | 2240 | ||
AY320284.2 | - | 19 | 2523 | |||
BC010663.1 | - | 16 | 1631 | |||
BC050351.1 | - | 19 | 2062 | 5264871 | ||
BC065220.1 | - | 19 | 2137 | 5732786 | ||
BC131591.1 | - | 19 | 2024 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
