siRNA Details
Gene Symbol: PACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100913.2 | NP_001094383.2 | 25 | 3406 | ||
NM_001243127.2 | NP_001230056.1 | 24 | 2883 | |||
NM_015197.3 | NP_056012.2 | 24 | 3361 | |||
XM_006720090.1 | XP_006720153.1 | 25 | 3074 | |||
XM_006720091.1 | XP_006720154.1 | 24 | 3041 | |||
XM_006720092.2 | XP_006720155.1 | 24 | 3002 | |||
XM_017021105.1 | XP_016876594.1 | 25 | 3062 | |||
XM_017021106.1 | XP_016876595.1 | 25 | 3059 | |||
XM_017021107.1 | XP_016876596.1 | 25 | 3050 | |||
XM_017021108.1 | XP_016876597.1 | 25 | 3047 | |||
XM_017021109.1 | XP_016876598.1 | 24 | 3038 | |||
XM_017021110.1 | XP_016876599.1 | 24 | 3029 | |||
XM_017021111.1 | XP_016876600.1 | 24 | 2907 | |||
XM_017021112.1 | XP_016876601.1 | 25 | 2842 | |||
GenBank mRNA | AB011174.1 | - | 25 | 3077 | ||
BC010663.1 | - | 21 | 2468 | |||
BC050351.1 | - | 24 | 2899 | 5264871 | ||
BC065220.1 | - | 24 | 2974 | 5732786 | ||
BC095493.1 | - | 1 | 811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
