siRNA Details
Gene Symbol: PACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100913.2 | NP_001094383.2 | 19, 20 | 2544 | ||
XM_006720090.1 | XP_006720153.1 | 19, 20 | 2212 | |||
XM_017021105.1 | XP_016876594.1 | 19, 20 | 2200 | |||
XM_017021106.1 | XP_016876595.1 | 19, 20 | 2197 | |||
XM_017021107.1 | XP_016876596.1 | 19, 20 | 2188 | |||
XM_017021108.1 | XP_016876597.1 | 19, 20 | 2185 | |||
XM_017021112.1 | XP_016876601.1 | 19, 20 | 1980 | |||
GenBank mRNA | AB011174.1 | - | 19, 20 | 2215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
