siRNA Details
Gene Symbol: PPBPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026769.1 | 1 | 791 | |||
GenBank mRNA | BC132938.1 | - | 1 | 714 | ||
BC132940.1 | - | 1 | 713 | |||
L10403.1 | - | 1 | 791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
