siRNA Details
Gene Symbol: ACBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039844.2 | NP_001034933.1 | 4 | 488 | ||
GenBank mRNA | AK095538.1 | - | 4 | 488 | ||
BC029526.1 | - | 4 | 488 | 5301753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
