siRNA Details
Gene Symbol: SPTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024858.2 | NP_001020029.1 | 35 | 10020 | ||
XM_005268023.4 | XP_005268080.1 | 36 | 10087 | |||
XM_017021612.1 | XP_016877101.1 | 37 | 10235 | |||
GenBank mRNA | AK094815.1 | - | 1 | 2163 | ||
BC010434.1 | - | 1 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
