siRNA Details
Gene Symbol: ZC3H12D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207360.2 | NP_997243.2 | 6 | 2242 | ||
GenBank mRNA | AB458221.1 | - | 6 | 2220 | ||
AB458222.1 | - | 7 | 1927 | |||
AK090441.1 | - | 1 | 2877 | |||
AK127932.1 | - | 7 | 1831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
