siRNA Details
Gene Symbol: IBA57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010867.3 | NP_001010867.1 | 2 | 674 | ||
NM_001310327.1 | NP_001297256.1 | 2 | 385 | |||
GenBank mRNA | AK022796.1 | - | 3 | 2701 | ||
AK303837.1 | - | 2 | 385 | |||
BI820993.1 | - | 2 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
