siRNA Details
Gene Symbol: SMIM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145432.1 | NP_001138904.1 | 3 | 750 | ||
GenBank mRNA | BC006003.1 | - | 3 | 694 | ||
BC032431.1 | - | 3 | 749 | |||
BC040154.1 | - | 3 | 655 | |||
BC046171.2 | - | 3 | 697 | |||
BC062747.1 | - | 3 | 680 | |||
BC086307.1 | - | 3 | 713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
