siRNA Details
Gene Symbol: FSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173651.3 | NP_775922.3 | 16 | 3971 | ||
GenBank mRNA | CR936597.1 | - | 11 | 3327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
