siRNA Details
Gene Symbol: ZSWIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242487.1 | NP_001229416.1 | 1 | 314 | ||
NM_001242488.1 | NP_001229417.1 | 1 | 314 | |||
NM_015037.3 | NP_055852.2 | 1 | 314 | |||
XM_005269648.2 | XP_005269705.1 | 1 | 361 | |||
XM_005269653.3 | XP_005269710.1 | 1 | 59 | |||
XM_005269655.3 | XP_005269712.1 | 1 | 59 | |||
XM_006717725.3 | XP_006717788.1 | 1 | 361 | |||
XM_011539544.1 | XP_011537846.1 | 1 | 361 | |||
XM_017015978.1 | XP_016871467.1 | 1 | 361 | |||
GenBank mRNA | AB020720.2 | - | 1 | 314 | ||
AK091621.1 | - | 1 | 361 | |||
BC137249.1 | - | 1 | 283 | |||
BC137250.1 | - | 1 | 283 | |||
BC151206.1 | - | 1 | 276 | 40147119 | ||
BM193415.1 | - | 1 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
