siRNA Details
Gene Symbol: EMC3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103821.1 | 3 | 1566 | |||
GenBank mRNA | AK092352.1 | - | 2 | 1514 | ||
AK125558.1 | - | 4 | 1335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
