siRNA Details
Gene Symbol: ERFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291832.1 | NP_001278761.1 | 8 | 1450 | ||
GenBank mRNA | AK094353.1 | - | 5 | 780 | ||
BC047423.1 | - | 4 | 793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
