siRNA Details
Gene Symbol: SRRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110199.2 | NP_001103669.1 | 7, 8 | 856 | ||
NM_001291831.1 | NP_001278760.1 | 7, 8 | 856 | |||
GenBank mRNA | BC029292.1 | - | 7, 8 | 849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
