siRNA Details
Gene Symbol: RIIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144956.1 | NP_001138428.1 | 4 | 279 | ||
GenBank mRNA | AA393535.1 | - | 4 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
