siRNA Details
Gene Symbol: C3orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168214.1 | NP_001161686.1 | 1 | 940 | ||
GenBank mRNA | BC031660.1 | - | 1 | 734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
