siRNA Details
Gene Symbol: FAM71E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145402.1 | NP_001138874.1 | 9 | 2696 | ||
GenBank mRNA | AK127979.1 | - | 8 | 2677 | ||
AL834316.1 | - | 2 | 1454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
