siRNA Details
Gene Symbol: REEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001330.2 | NP_001001330.1 | 8 | 3933 | ||
GenBank mRNA | BC018658.1 | - | 1 | 451 | ||
CR936686.1 | - | 1 | 1667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
