siRNA Details
Gene Symbol: ZNF609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015042.1 | NP_055857.1 | 4 | 2665 | ||
XM_011521386.2 | XP_011519688.1 | 5 | 2891 | |||
XM_017022021.1 | XP_016877510.1 | 5 | 2873 | |||
GenBank mRNA | AB002293.1 | - | 1 | 1376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
